Assay Details
Target Gene Details
Entrez Gene ID: | 677 |
Gene Name: | ZFP36 ring finger protein like 1 |
Gene Aliases: |
BRF1, Berg36, ERF-1, ERF1, RNF162B, TIS11B, cMG1 |
Location: |
Chr.14:68787655-68796243 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFP36L1 | NM_001244698.1 | NP_001231627.1 | ||
| NM_001244701.1 | NP_001231630.1 | |||
| NM_004926.3 | NP_004917.2 | |||
| AK024202.1 | ||||
| AK294363.1 | ||||
| AK303917.1 | ||||
| BC018340.1 | AAH18340.1 | |||
| BP353132.1 | ||||
| BT019468.1 | AAV38275.1 | |||
| DA633985.1 | ||||
| FY210721.1 | ||||
| X79066.1 | 1 | 663 | CAA55670.1 | |
| X99404.1 | CAA67781.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070226 | Chr.14:68788582 - 68795483 on Build GRCh38 | Deletion |
|
| nsv952859 | Chr.14:68788584 - 68795783 on Build GRCh38 | Deletion |
|
| dgv1097n106 | Chr.14:68790783 - 68795883 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map