Assay Details
Target Gene Details
Entrez Gene ID: | 162417 |
Gene Name: | N-acetylglutamate synthase |
Gene Aliases: |
AGAS, ARGA |
Location: |
Chr.17:44004546-44009068 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAGS | NM_153006.2 | 7 | 1957 | NP_694551.1 |
| XM_011524438.1 | 6 | 1892 | XP_011522740.1 | |
| XM_011524439.2 | 7 | 1847 | XP_011522741.1 | |
| AI241153.1 | 1 | 130 | ||
| AK074407.1 | 7 | 1887 | ||
| AK091938.1 | 4 | 1845 | ||
| BF589712.1 | 1 | 115 |
Target Gene Details
Entrez Gene ID: | 5697 |
Gene Name: | peptide YY |
Gene Aliases: |
PYY-I, PYY1 |
Location: |
Chr.17:43952733-44024260 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PYY | XM_011525035.1 | XP_011523337.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833454 | Chr.17:43940848 - 44158311 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map