Assay Details
Target Gene Details
Entrez Gene ID: | 5589 |
Gene Name: | protein kinase C substrate 80K-H |
Gene Aliases: |
AGE-R2, G19P1, GIIB, PCLD, PKCSH, PLD1, VASAP-60 |
Location: |
Chr.19:11435257-11450968 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKCSH | NM_001001329.2 | 6 | 930 | NP_001001329.1 |
| NM_001289102.1 | 5 | 781 | NP_001276031.1 | |
| NM_001289103.1 | 6 | 934 | NP_001276032.1 | |
| NM_001289104.1 | 6 | 930 | NP_001276033.1 | |
| NM_002743.3 | 6 | 542 | NP_002734.2 | |
| XM_011528130.1 | 7 | 696 | XP_011526432.1 | |
| XM_011528131.1 | 7 | 696 | XP_011526433.1 | |
| XM_011528132.1 | 7 | 696 | XP_011526434.1 | |
| XM_017026977.1 | 6 | 546 | XP_016882466.1 | |
| AF144075.1 | 5 | 403 | AAF66686.1 | |
| AK130663.1 | 6 | 558 | ||
| AK225279.1 | 6 | 556 | ||
| AK225390.1 | 6 | 538 | ||
| AK290433.1 | 5 | 776 | ||
| AK296187.1 | 6 | 557 | ||
| BC013586.1 | 5 | 340 | ||
| BC128412.1 | 2 | 142 | ||
| BF984334.1 | 2 | 94 | ||
| BT009858.1 | AAP88860.1 | |||
| J03075.1 | 6 | 539 | AAA52493.1 | |
| KU178278.1 | ||||
| KU178279.1 | ||||
| KU178280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833752 | Chr.19:11208324 - 11448939 on Build GRCh38 | Loss |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map