Assay Details
Target Gene Details
Entrez Gene ID: | 10942 |
Gene Name: | protease, serine 21 |
Gene Aliases: |
ESP-1, ESP1, TEST1, TESTISIN |
Location: |
Chr.16:2817163-2821722 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS21 | NM_001270452.1 | NP_001257381.1 | ||
| NM_006799.3 | NP_006790.1 | |||
| NM_144956.2 | NP_659205.1 | |||
| NM_144957.2 | NP_659206.1 | |||
| NR_073012.1 | ||||
| XM_017022875.1 | XP_016878364.1 | |||
| XM_017022876.1 | XP_016878365.1 | |||
| XM_017022877.1 | XP_016878366.1 | |||
| XM_017022878.1 | XP_016878367.1 | |||
| AB031329.1 | BAA83520.1 | |||
| AB031330.1 | BAA83521.1 | |||
| AB031331.1 | BAA89532.1 | |||
| AF058300.1 | AAD41588.1 | |||
| AY359034.1 | AAQ89393.1 | |||
| BC074999.2 | AAH74999.1 | |||
| BC075000.2 | AAH75000.1 | |||
| BG717268.1 | ||||
| BI334059.1 | ||||
| BI827608.1 | ||||
| BX454526.1 | ||||
| CB529999.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952905 | Chr.16:2817200 - 2863199 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79724175] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map