Assay Details
Target Gene Details
Entrez Gene ID: | 257240 |
Gene Name: | kelch like family member 34 |
Gene Aliases: |
- |
Location: |
Chr.X:21655349-21658387 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL34 | NM_153270.2 | 1 | 989 | NP_695002.1 |
| AK092279.1 | 1 | 932 | BAC03848.1 | |
| BC109031.2 | 1 | 901 | AAI09032.1 |
More Information
Set Membership: |
|

Genomic Map