Assay Details
Target Gene Details
Entrez Gene ID: | 54509 |
Gene Name: | ras homolog family member F, filopodia associated |
Gene Aliases: |
ARHF, RIF |
Location: |
Chr.12:121777754-121793688 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOF | NM_019034.2 | 5 | 1576 | NP_061907.2 |
| AK000254.1 | 5 | 1542 | BAA91034.1 | |
| BM830620.1 | 1 | 217 | ||
| BQ422695.1 |
Target Gene Details
Entrez Gene ID: | 144404 |
Gene Name: | transmembrane protein 120B |
Gene Aliases: |
- |
Location: |
Chr.12:121712752-121782070 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM120B | NM_001080825.2 | 12 | 4055 | NP_001074294.2 |
| XM_005253843.4 | XP_005253900.1 | |||
| XM_017018853.1 | XP_016874342.1 | |||
| XM_017018854.1 | XP_016874343.1 | |||
| AK123181.1 | ||||
| BX648084.1 | 8 | 3528 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892268 | Chr.12:121747658 - 121845875 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map