Assay Details
Target Gene Details
Entrez Gene ID: | 9121 |
Gene Name: | solute carrier family 16 member 5 |
Gene Aliases: |
MCT5, MCT6 |
Location: |
Chr.17:75087727-75106590 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A5 | NM_001271765.1 | 2 | 257 | NP_001258694.1 |
| NM_004695.3 | 2 | 191 | NP_004686.1 | |
| XM_017025293.1 | 2 | 169 | XP_016880782.1 | |
| AK000416.1 | 2 | 257 | ||
| AK092512.1 | 1 | 300 | ||
| BC009684.1 | 2 | 172 | AAH09684.1 | |
| BC033611.1 | 1 | 81 | AAH33611.1 | |
| BI820515.1 | 2 | 191 |
More Information
Set Membership: |
|

Genomic Map