Assay Details
Target Gene Details
Entrez Gene ID: | 1043 |
Gene Name: | CD52 molecule |
Gene Aliases: |
CDW52 |
Location: |
Chr.1:26317920-26320523 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD52 | NM_001803.2 | 1 | 89 | NP_001794.2 |
| A23013.1 | CAA01644.1 | |||
| AK311554.1 | 1 | 80 | ||
| BC000644.2 | AAH00644.1 | |||
| BI835519.1 | 1 | 91 | ||
| BT007116.1 | AAP35780.1 | |||
| CR456874.1 | CAG33155.1 | |||
| FJ460513.1 | 1 | 89 | ||
| KP176645.1 | ||||
| X62466.1 | CAA44323.1 | |||
| X67699.1 | CAA47929.1 |
Target Gene Details
Entrez Gene ID: | 91544 |
Gene Name: | UBX domain protein 11 |
Gene Aliases: |
COA-1, PP2243, SOC, SOCI, UBXD5 |
Location: |
Chr.1:26282282-26318265 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN11 | NM_145345.2 | NP_663320.2 | ||
| AK097368.1 | BAC05024.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77928789] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map