Assay Details
Target Gene Details
Entrez Gene ID: | 151050 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 like |
Gene Aliases: |
C2orf67, MSL1v2 |
Location: |
Chr.2:210021421-210171370 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KANSL1L | NM_001307976.1 | 14 | 3829 | NP_001294905.1 |
| NM_152519.3 | 15 | 3955 | NP_689732.2 | |
| XM_005246328.4 | 15 | 4237 | XP_005246385.1 | |
| XM_005246329.4 | 15 | 3943 | XP_005246386.1 | |
| XM_005246330.4 | 15 | 3824 | XP_005246387.1 | |
| XM_005246332.4 | 14 | 4110 | XP_005246389.1 | |
| XM_011510706.2 | 15 | 3862 | XP_011509008.1 | |
| XM_011510707.2 | 15 | 4045 | XP_011509009.1 | |
| XM_011510710.2 | 14 | 2694 | XP_011509012.1 | |
| XM_017003430.1 | 15 | 3981 | XP_016858919.1 | |
| XM_017003431.1 | 11 | 6063 | XP_016858920.1 | |
| AA252480.1 | 1 | 108 | ||
| AA404520.1 | 1 | 60 | ||
| AK074441.1 | 14 | 2724 | BAB85085.1 | |
| BQ948513.1 | 1 | 501 | ||
| CA393360.1 |
Target Gene Details
Entrez Gene ID: | 6120 |
Gene Name: | ribulose-5-phosphate-3-epimerase |
Gene Aliases: |
RPE2-1 |
Location: |
Chr.2:210002565-210029156 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPE | NM_001278282.1 | 7 | 3207 | NP_001265211.1 |
| NM_001278283.1 | 8 | 3287 | NP_001265212.1 | |
| NM_001278285.1 | 5 | 3115 | NP_001265214.1 | |
| NM_001278286.1 | 6 | 3131 | NP_001265215.1 | |
| NM_001278288.1 | 5 | 3122 | NP_001265217.1 | |
| NM_001278289.1 | 7 | 3271 | NP_001265218.1 | |
| NM_001318926.1 | 7 | 3256 | NP_001305855.1 | |
| NM_001318927.1 | 7 | 3300 | NP_001305856.1 | |
| NM_001318928.1 | 7 | 3151 | NP_001305857.1 | |
| NM_001318929.1 | 6 | 3198 | NP_001305858.1 | |
| NM_001318930.1 | 6 | 3191 | NP_001305859.1 | |
| NM_001318931.1 | 6 | 3153 | NP_001305860.1 | |
| NM_006916.2 | 6 | 3176 | NP_008847.1 | |
| NM_199229.2 | 6 | 3202 | NP_954699.1 | |
| XM_006712677.3 | 7 | 3149 | XP_006712740.1 | |
| AB075523.1 | 1 | 1827 | ||
| AI222711.1 | 1 | 64 |
More Information
Set Membership: |
|

Genomic Map