Assay Details
Target Gene Details
Entrez Gene ID: | 29088 |
Gene Name: | mitochondrial ribosomal protein L15 |
Gene Aliases: |
HSPC145, L15mt, MRP-L15, MRP-L7, RPML7 |
Location: |
Chr.8:54135210-54148514 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL15 | NM_014175.3 | NP_054894.1 | ||
| XM_017013362.1 | XP_016868851.1 | |||
| AF161494.1 | AAF29109.1 | |||
| AK312832.1 | ||||
| AL136665.1 | CAB66600.1 | |||
| BC000891.1 | AAH00891.1 | |||
| CR533531.1 | CAG38562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv820132 | Chr.8:54135203 - 54135673 on Build GRCh38 | Gain |
|
| nsv1026869 | Chr.8:54077172 - 54176800 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map