Assay Details
Target Gene Details
Entrez Gene ID: | 140901 |
Gene Name: | serine/threonine kinase 35 |
Gene Aliases: |
CLIK1, STK35L1 |
Location: |
Chr.20:2098035-2148555 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK35 | NM_080836.3 | 2 | 461 | NP_543026.2 |
| XM_011529174.2 | 2 | 4308 | XP_011527476.1 | |
| AF195026.1 | 1 | 62 | AAL99353.1 | |
| AK302629.1 | ||||
| AK314728.1 | 2 | 313 | ||
| BC017340.2 | 1 | 44 | AAH17340.2 | |
| BC111573.1 | AAI11574.1 | |||
| BC140883.1 | 1 | 162 | ||
| BC140884.1 | 1 | 162 | ||
| GQ281297.1 | 2 | 461 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| nsv3258 | Chr.20:2091289 - 2124098 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map