Assay Details
Target Gene Details
Entrez Gene ID: | 2976 |
Gene Name: | general transcription factor IIIC subunit 2 |
Gene Aliases: |
TFIIIC-BETA, TFIIIC110 |
Location: |
Chr.2:27325849-27357034 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3C2 | NM_001035521.2 | 19 | 3498 | NP_001030598.1 |
| NM_001318909.1 | 19 | 3608 | NP_001305838.1 | |
| NM_001521.3 | 20 | 3574 | NP_001512.1 | |
| XM_005264272.4 | 20 | 3782 | XP_005264329.1 | |
| XM_011532801.2 | 20 | 3921 | XP_011531103.1 | |
| AF054988.1 | 7 | 1323 | AAC09349.1 | |
| BC020981.1 | 18 | 3207 | AAH20981.1 | |
| BU632224.1 | 1 | 403 | ||
| D13636.1 | 18 | 3217 | BAA02800.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
| nsv2654 | Chr.2:27309443 - 27336770 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map