Assay Details
Target Gene Details
Entrez Gene ID: | 9815 |
Gene Name: | GIT ArfGAP 2 |
Gene Aliases: |
CAT-2, CAT2, PKL |
Location: |
Chr.12:109929792-109996389 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIT2 | NM_001135213.1 | 19 | 4565 | NP_001128685.1 |
| NM_001135214.1 | 19 | 4709 | NP_001128686.1 | |
| NM_014776.3 | 18 | 4559 | NP_055591.2 | |
| NM_057169.3 | 20 | 4799 | NP_476510.1 | |
| NM_057170.3 | 18 | 4415 | NP_476511.1 | |
| XM_005253997.3 | 20 | 4791 | XP_005254054.1 | |
| XM_005254000.3 | 19 | 4641 | XP_005254057.1 | |
| XM_006719707.3 | 20 | 4749 | XP_006719770.1 | |
| XM_006719708.3 | 19 | 4659 | XP_006719771.1 | |
| XM_006719709.3 | 19 | 4644 | XP_006719772.1 | |
| XM_006719712.3 | 17 | 4305 | XP_006719775.1 | |
| XM_017020258.1 | 19 | 4656 | XP_016875747.1 | |
| XM_017020259.1 | 20 | 4795 | XP_016875748.1 | |
| AK057814.1 | 1 | 321 | ||
| AK090652.1 | 1 | 2044 | ||
| AK091325.1 | 1 | 1511 | ||
| CR749340.1 | 6 | 3290 | CAH18193.1 | |
| D63482.2 | 19 | 4471 |
More Information
Set Membership: |
|

Genomic Map