Assay Details
Target Gene Details
Entrez Gene ID: | 26277 |
Gene Name: | TERF1 interacting nuclear factor 2 |
Gene Aliases: |
DKCA3, TIN2 |
Location: |
Chr.14:24239641-24242674 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TINF2 | NM_001099274.1 | NP_001092744.1 | ||
| NM_012461.2 | NP_036593.2 | |||
| XM_005267529.3 | XP_005267586.1 | |||
| XM_011536642.2 | XP_011534944.1 | |||
| XM_017021216.1 | XP_016876705.1 | |||
| XM_017021217.1 | XP_016876706.1 | |||
| AF195512.1 | AAF18439.1 | |||
| AK023166.1 | 3 | 1252 | BAB14440.1 | |
| AK225379.1 | ||||
| AK294119.1 | ||||
| AK315084.1 | ||||
| BC005030.1 | AAH05030.1 | |||
| BC019343.1 | AAH19343.1 | |||
| BM549897.1 | ||||
| BX161478.1 | CAD61933.1 | |||
| EU851975.1 |
More Information
Set Membership: |
|

Genomic Map