Assay Details
Target Gene Details
Entrez Gene ID: | 127253 |
Gene Name: | tRNA-yW synthesizing protein 3 homolog |
Gene Aliases: |
C1orf171 |
Location: |
Chr.1:74733143-74766677 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TYW3 | NM_001162916.1 | 5 | 1483 | NP_001156388.1 |
| NM_138467.2 | 6 | 1578 | NP_612476.1 | |
| NR_027962.1 | 6 | 1669 | ||
| XM_006710347.1 | XP_006710410.1 | |||
| AK098237.1 | 6 | 1578 | BAC05267.1 | |
| BC071765.1 | 6 | 1578 | AAH71765.1 | |
| BX647591.1 | 2 | 1025 | CAI46106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830259 | Chr.1:74623493 - 74775378 on Build GRCh38 | Gain |
|
| nsv1002115 | Chr.1:72379872 - 74933684 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map