Assay Details
Target Gene Details
Entrez Gene ID: | 23527 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 2 |
Gene Aliases: |
CENTB2, CNT-B2 |
Location: |
Chr.3:195274736-195443176 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAP2 | NM_012287.5 | 23 | 6642 | NP_036419.3 |
| XM_006713557.3 | XP_006713620.1 | |||
| XM_017006046.1 | 26 | 6876 | XP_016861535.1 | |
| XM_017006048.1 | XP_016861537.1 | |||
| XM_017006049.1 | XP_016861538.1 | |||
| BC060767.1 | 23 | 6577 | AAH60767.1 | |
| CA413842.1 | 1 | 546 | ||
| D26069.3 | 23 | 6411 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599226 | Chr.3:194987301 - 195510679 on Build GRCh38 | Gain |
|
| nsv1009796 | Chr.3:195016604 - 195403519 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map