Assay Details
Target Gene Details
Entrez Gene ID: | 284904 |
Gene Name: | SEC14 like lipid binding 4 |
Gene Aliases: |
TAP3 |
Location: |
Chr.22:30488150-30505711 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC14L4 | NM_001161368.1 | 12 | 2026 | NP_001154840.1 |
| NM_174977.3 | 12 | 2394 | NP_777637.1 | |
| XM_006724235.2 | 12 | 2410 | XP_006724298.1 | |
| XM_011530164.2 | 12 | 2574 | XP_011528466.1 | |
| XM_017028787.1 | 11 | 2523 | XP_016884276.1 | |
| XM_017028788.1 | 10 | 2440 | XP_016884277.1 | |
| XM_017028789.1 | 11 | 2196 | XP_016884278.1 | |
| XM_017028791.1 | 11 | 2223 | XP_016884280.1 | |
| AK092002.1 | 13 | 2443 | ||
| AK093418.1 | 12 | 2455 | ||
| AK093448.1 | 12 | 2378 | ||
| T72380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map