Assay Details
Target Gene Details
Entrez Gene ID: | 51285 |
Gene Name: | RAS like family 12 |
Gene Aliases: |
RIS |
Location: |
Chr.15:65045576-65076654 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASL12 | NM_001307930.1 | 4 | 1661 | NP_001294859.1 |
| NM_016563.3 | 5 | 1718 | NP_057647.1 | |
| XM_005254434.4 | XP_005254491.1 | |||
| XM_011521660.2 | 6 | 1531 | XP_011519962.1 | |
| XM_017022296.1 | XP_016877785.1 | |||
| AF233588.1 | 5 | 1557 | AAF60286.1 | |
| BC053734.1 | 5 | 1566 | AAH53734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763104 | Chr.15:64923655 - 65128399 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map