Assay Details
Target Gene Details
Entrez Gene ID: | 55929 |
Gene Name: | DNA methyltransferase 1 associated protein 1 |
Gene Aliases: |
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4 |
Location: |
Chr.1:44213428-44220679 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DMAP1 | NM_001034023.1 | 7 | 888 | NP_001029195.1 |
| NM_001034024.1 | 7 | 848 | NP_001029196.1 | |
| NM_019100.4 | 6 | 1069 | NP_061973.1 | |
| XM_005271037.4 | 6 | 1059 | XP_005271094.1 | |
| XM_005271039.1 | 7 | 959 | XP_005271096.1 | |
| XM_005271040.1 | 7 | 836 | XP_005271097.1 | |
| XM_006710771.3 | 5 | 738 | XP_006710834.1 | |
| XM_011541784.2 | 7 | 893 | XP_011540086.1 | |
| XM_011541785.2 | 7 | 913 | XP_011540087.1 | |
| XM_017001807.1 | 7 | 944 | XP_016857296.1 | |
| XM_017001808.1 | 7 | 947 | XP_016857297.1 | |
| AB037846.1 | 7 | 853 | ||
| AF265228.1 | 6 | 768 | AAF87079.1 | |
| AK021605.1 | 7 | 832 | BAB13854.1 | |
| AK054954.1 | 3 | 1686 | ||
| AK092898.1 | 3 | 3853 | ||
| AK096252.1 | 3 | 2456 | ||
| AK289366.1 | 7 | 871 | ||
| AK298793.1 | 7 | 831 | ||
| AL136657.1 | 7 | 850 | CAB66592.1 | |
| AL137200.1 | 7 | 852 | CAB69910.1 | |
| BC002855.2 | 7 | 837 | AAH02855.1 | |
| BC008053.2 | 6 | 1031 | AAH08053.1 | |
| BX537895.1 | 3 | 3007 | CAD97886.1 |
More Information
Set Membership: |
|

Genomic Map