Assay Details
Target Gene Details
Entrez Gene ID: | 2531 |
Gene Name: | 3-ketodihydrosphingosine reductase |
Gene Aliases: |
DHSR, FVT1, SDR35C1 |
Location: |
Chr.18:63327736-63367273 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDSR | NM_002035.2 | NP_002026.1 | ||
| XM_005266677.2 | XP_005266734.1 | |||
| XM_017025690.1 | XP_016881179.1 | |||
| AK025120.1 | ||||
| AK126679.1 | ||||
| AK297670.1 | ||||
| AK312360.1 | ||||
| BT006782.1 | AAP35428.1 | |||
| X63657.1 | CAA45197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv577096 | Chr.18:63311353 - 63347135 on Build GRCh38 | Loss |
|
| nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map