Assay Details
Target Gene Details
Entrez Gene ID: | 2897 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 1 |
Gene Aliases: |
EAA3, EEA3, GLR5, GLUR5, GluK1, gluR-5 |
Location: |
Chr.21:29536933-29940052 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK1 | NM_001320616.1 | 17 | 3313 | NP_001307545.1 |
| NM_001320618.1 | 14 | 2896 | NP_001307547.1 | |
| NM_001320621.1 | 14 | 2842 | NP_001307550.1 | |
| NM_175611.2 | 16 | 3180 | NP_783300.1 | |
| XM_005260942.1 | 17 | 2973 | XP_005260999.1 | |
| XM_006723991.1 | 18 | 3018 | XP_006724054.1 | |
| AA404271.1 | 1 | 310 | ||
| AJ249208.1 | CAC80546.1 | |||
| AK296404.1 | 17 | 3258 | ||
| U16125.1 | AAA95961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834073 | Chr.21:29386820 - 29553837 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map