Assay Details
Target Gene Details
Entrez Gene ID: | 84265 |
Gene Name: | RNA polymerase III subunit G like |
Gene Aliases: |
RPC32HOM, flj32422 |
Location: |
Chr.1:145959037-145978848 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR3GL | NM_032305.1 | NP_115681.1 | ||
| XM_005277431.4 | XP_005277488.1 | |||
| XM_011510059.2 | XP_011508361.1 | |||
| AK056984.1 | BAB71332.1 | |||
| AK092209.1 | ||||
| BC004355.1 | AAH04355.1 | |||
| DQ418461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005055 | Chr.1:145642988 - 148991068 on Build GRCh38 | Loss |
|
| nsv509457 | Chr.1:145948024 - 149021821 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map