Assay Details
Target Gene Details
Entrez Gene ID: | 56899 |
Gene Name: | ankyrin repeat and sterile alpha motif domain containing 1B |
Gene Aliases: |
AIDA, AIDA-1, ANKS2, EB-1, EB1, cajalin-2 |
Location: |
Chr.12:98729904-99984727 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Target Gene Details
Entrez Gene ID: | 196472 |
Gene Name: | family with sequence similarity 71 member C |
Gene Aliases: |
- |
Location: |
Chr.12:99647750-99650114 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71C | NM_153364.3 | 2 | 1269 | NP_699195.1 |
| BC031221.1 | 2 | 1266 | AAH31221.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32717 | Chr.12:99583091 - 99688529 on Build GRCh38 | Gain |
|
| nsv559954 | Chr.12:99496462 - 99657225 on Build GRCh38 | Loss |
|
| dgv1549n100 | Chr.12:99576473 - 99758416 on Build GRCh38 | Loss |
|
| nsv559956 | Chr.12:99619390 - 99795476 on Build GRCh38 | Loss |
|
| nsv1045504 | Chr.12:99626949 - 99720643 on Build GRCh38 | Loss |
|
| dgv2824n54 | Chr.12:99647996 - 99703568 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map