Assay Details
Target Gene Details
Entrez Gene ID: | 53632 |
Gene Name: | protein kinase AMP-activated non-catalytic subunit gamma 3 |
Gene Aliases: |
AMPKG3 |
Location: |
Chr.2:218821853-218831803 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAG3 | NM_017431.2 | 14 | 2079 | NP_059127.2 |
| XM_017004343.1 | 13 | 2662 | XP_016859832.1 | |
| AA256383.1 | 1 | 225 | ||
| AF214519.1 | 12 | 1978 | AAF73987.1 | |
| AJ249977.1 | 14 | 2080 | CAB65117.1 |
More Information
Set Membership: |
|

Genomic Map