Assay Details
Target Gene Details
Entrez Gene ID: | 54890 |
Gene Name: | alkB homolog 5, RNA demethylase |
Gene Aliases: |
ABH5, OFOXD, OFOXD1 |
Location: |
Chr.17:18183553-18209954 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALKBH5 | NM_017758.3 | 3 | 1597 | NP_060228.3 |
| AK000129.1 | 1 | 45 | ||
| AK000315.1 | 3 | 948 | BAA91078.1 | |
| AK301107.1 | 4 | 980 | ||
| AK311161.1 | 3 | 324 | ||
| AK311511.1 | 3 | 289 | ||
| AL137263.1 | 1 | 309 | ||
| BC062339.1 | 3 | 1175 | AAH62339.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510702 | Chr.17:18134362 - 18213487 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map