Assay Details
Target Gene Details
Entrez Gene ID: | 3728 |
Gene Name: | junction plakoglobin |
Gene Aliases: |
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB |
Location: |
Chr.17:41754607-41786853 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JUP | NM_002230.2 | NP_002221.1 | ||
NM_021991.2 | NP_068831.1 | |||
XM_006721873.2 | XP_006721936.1 | |||
XM_006721874.2 | XP_006721937.1 | |||
XM_006721875.1 | XP_006721938.1 | |||
XM_006721878.1 | XP_006721941.1 | |||
XM_011524753.2 | XP_011523055.1 | |||
XM_011524755.1 | XP_011523057.1 | |||
XM_011524756.1 | XP_011523058.1 | |||
XM_011524757.2 | XP_011523059.1 | |||
XM_011524758.1 | XP_011523060.1 | |||
XM_017024588.1 | XP_016880077.1 | |||
XM_017024589.1 | XP_016880078.1 | |||
XM_017024590.1 | XP_016880079.1 | |||
AK226051.1 | ||||
AY243535.1 | AAO85780.1 | |||
BC000441.2 | AAH00441.2 | |||
BC011865.2 | AAH11865.1 | |||
KU178155.1 | ||||
KU178156.1 | ||||
M23410.1 | AAA64895.1 | |||
Z68228.1 | CAA92522.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
