Assay Details
Target Gene Details
Entrez Gene ID: | 54716 |
Gene Name: | solute carrier family 6 member 20 |
Gene Aliases: |
SIT1, XT3, Xtrp3 |
Location: |
Chr.3:45755449-45796553 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A20 | NM_020208.3 | 11 | 1806 | NP_064593.1 |
| NM_022405.3 | 10 | 1695 | NP_071800.1 | |
| XM_011533847.2 | 10 | 2120 | XP_011532149.1 | |
| AF125107.1 | 12 | 1870 | AAL75944.1 | |
| AJ276207.1 | 11 | 1733 | CAB99310.1 | |
| AJ276208.1 | 10 | 1622 | CAB99311.1 | |
| AL389979.1 | 6 | 1870 | CAB97535.1 | |
| BC126197.1 | 11 | 1746 | ||
| BC136431.1 | 11 | 1746 | ||
| BC144087.1 |
More Information
Set Membership: |
|

Genomic Map