Assay Details
Target Gene Details
Entrez Gene ID: | 4112 |
Gene Name: | MAGE family member B1 |
Gene Aliases: |
CT3.1, DAM10, MAGE-Xp, MAGEL1 |
Location: |
Chr.X:30243731-30252038 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB1 | NM_002363.4 | NP_002354.2 | ||
| NM_177404.2 | NP_796379.1 | |||
| NM_177415.2 | NP_803134.1 | |||
| AK314977.1 | ||||
| BC013772.1 | AAH13772.1 | |||
| BU166425.1 | ||||
| DB022312.1 | ||||
| X82539.1 | CAA57889.1 |
More Information
Set Membership: |
|

Genomic Map