Assay Details
Target Gene Details
Entrez Gene ID: | 199731 |
Gene Name: | cell adhesion molecule 4 |
Gene Aliases: |
IGSF4C, NECL4, Necl-4, TSLL2, synCAM4 |
Location: |
Chr.19:43622368-43641984 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CADM4 | NM_145296.1 | NP_660339.1 | ||
| XM_005258620.2 | XP_005258677.1 | |||
| XM_017026452.1 | XP_016881941.1 | |||
| AF363368.1 | AAM60750.1 | |||
| AK313028.1 | ||||
| BC113963.1 | AAI13964.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833839 | Chr.19:43595661 - 43768147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map