Assay Details
Target Gene Details
Entrez Gene ID: | 1891 |
Gene Name: | enoyl-CoA hydratase 1 |
Gene Aliases: |
HPXEL |
Location: |
Chr.19:38815422-38831857 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECH1 | NM_001398.2 | 10 | 1026 | NP_001389.2 |
| XM_017026448.1 | 10 | 976 | XP_016881937.1 | |
| AK126566.1 | 7 | 3425 | ||
| AK291860.1 | 10 | 961 | ||
| AV738478.1 | 2 | 198 | ||
| BC011792.2 | 10 | 943 | AAH11792.1 | |
| BC017408.1 | 10 | 967 | AAH17408.1 | |
| U16660.1 | 10 | 968 | AAC50222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9722 | Chr.19:38807433 - 38816933 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map