Assay Details
Target Gene Details
Entrez Gene ID: | 50700 |
Gene Name: | retinol dehydrogenase 8 (all-trans) |
Gene Aliases: |
PRRDH, SDR28C2 |
Location: |
Chr.19:10013249-10022278 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDH8 | NM_015725.2 | 5 | 901 | NP_056540.2 |
| AF229845.1 | 5 | 901 | AAF63160.1 | |
| AK024022.1 | 5 | 666 | BAB14782.1 |
More Information
Set Membership: |
|

Genomic Map