Assay Details
Target Gene Details
Entrez Gene ID: | 6136 |
Gene Name: | ribosomal protein L12 |
Gene Aliases: |
L12 |
Location: |
Chr.9:127447674-127451432 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPL12 | NM_000976.3 | NP_000967.1 | ||
| AF037643.1 | ||||
| AK057602.1 | ||||
| CB155565.1 | ||||
| L06505.1 | AAA36157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv972452 | Chr.9:127446981 - 127450055 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map