Assay Details
Target Gene Details
Entrez Gene ID: | 5174 |
Gene Name: | PDZ domain containing 1 |
Gene Aliases: |
CAP70, CLAMP, NHERF-3, NHERF3, PDZD1 |
Location: |
Chr.1:145670852-145707507 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDZK1 | NM_001201325.1 | NP_001188254.1 | ||
| NM_001201326.1 | NP_001188255.1 | |||
| NM_002614.4 | NP_002605.2 | |||
| XM_011509616.2 | XP_011507918.1 | |||
| XM_011509618.2 | XP_011507920.1 | |||
| XM_011509619.2 | XP_011507921.2 | |||
| XM_017001464.1 | XP_016856953.1 | |||
| XM_017001465.1 | XP_016856954.1 | |||
| XM_017001466.1 | XP_016856955.1 | |||
| XM_017001467.1 | XP_016856956.1 | |||
| AF012281.1 | AAC12264.1 | |||
| AK298269.1 | ||||
| AK313100.1 | ||||
| BC006496.2 | AAH06496.1 | |||
| BC006518.2 | AAH06518.1 | |||
| BX477678.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587421 | Chr.1:145649153 - 145751059 on Build GRCh38 | Loss |
|
| dgv331n100 | Chr.1:145642988 - 145827134 on Build GRCh38 | Gain |
|
| esv3572342 | Chr.1:145601946 - 145809476 on Build GRCh38 | Gain |
|
| nsv469752 | Chr.1:145639438 - 145790042 on Build GRCh38 | Loss |
|
| nsv831392 | Chr.1:145632374 - 145813165 on Build GRCh38 | Gain |
|
| esv3894089 | Chr.1:145672100 - 145824903 on Build GRCh38 | Loss |
|
| dgv532n54 | Chr.1:145624561 - 145813526 on Build GRCh38 | Gain |
|
| dgv33n27 | Chr.1:145687602 - 145824903 on Build GRCh38 | Loss |
|
| dgv95e212 | Chr.1:145631218 - 145812561 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map