Assay Details
Target Gene Details
Entrez Gene ID: | 343099 |
Gene Name: | coiled-coil domain containing 18 |
Gene Aliases: |
NY-SAR-41 |
Location: |
Chr.1:93179802-93279037 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830625 | Chr.1:93100821 - 93269689 on Build GRCh38 | Loss |
|
| nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map