Assay Details
Target Gene Details
Entrez Gene ID: | 56895 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 4 |
Gene Aliases: |
1-AGPAT4, LPAAT-delta, dJ473J16.2 |
Location: |
Chr.6:161129980-161274108 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGPAT4 | NM_020133.2 | 9 | 5246 | NP_064518.1 |
| XM_005267053.3 | 8 | 5062 | XP_005267110.1 | |
| XM_006715514.3 | 8 | 5070 | XP_006715577.1 | |
| XM_017011059.1 | 10 | 5416 | XP_016866548.1 | |
| AL122123.1 | 1 | 2643 | ||
| CR627395.1 | 8 | 5027 | CAH10489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3611453 | Chr.6:160930783 - 161147021 on Build GRCh38 | Gain |
|
| esv3576242 | Chr.6:160924181 - 161141222 on Build GRCh38 | Gain |
|
| nsv1030045 | Chr.6:160938043 - 161141223 on Build GRCh38 | Gain |
|
| nsv1026543 | Chr.6:160306836 - 161585804 on Build GRCh38 | Gain |
|
| nsv1030852 | Chr.6:161056718 - 161168362 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map