Assay Details
Target Gene Details
Entrez Gene ID: | 148932 |
Gene Name: | MOB kinase activator 3C |
Gene Aliases: |
MOB1E, MOBKL2C |
Location: |
Chr.1:46607715-46616891 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOB3C | NM_201403.2 | NP_958805.1 | ||
| BI601393.1 | ||||
| CF125162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009193 | Chr.1:46474327 - 46619995 on Build GRCh38 | Gain |
|
| nsv522319 | Chr.1:46615069 - 46628595 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map