Assay Details
Target Gene Details
Entrez Gene ID: | 80333 |
Gene Name: | potassium voltage-gated channel interacting protein 4 |
Gene Aliases: |
CALP, KCHIP4 |
Location: |
Chr.4:20728520-21948801 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNIP4 | NM_001035003.1 | NP_001030175.1 | ||
| NM_001035004.1 | NP_001030176.1 | |||
| NM_025221.5 | NP_079497.2 | |||
| NM_147181.3 | NP_671710.1 | |||
| NM_147182.3 | NP_671711.1 | |||
| NM_147183.3 | NP_671712.1 | |||
| XM_005248190.4 | XP_005248247.1 | |||
| XM_011513882.2 | 8 | 2723 | XP_011512184.1 | |
| XM_011513883.2 | 7 | 2592 | XP_011512185.1 | |
| XM_011513884.2 | 7 | 2616 | XP_011512186.1 | |
| XM_011513885.2 | 7 | 7912 | XP_011512187.1 | |
| XM_011513886.2 | 7 | 2627 | XP_011512188.1 | |
| XM_011513887.2 | 7 | 3392 | XP_011512189.1 | |
| XM_011513889.2 | 7 | 2582 | XP_011512191.1 | |
| XM_017008653.1 | XP_016864142.1 | |||
| AF302044.1 | AAG36974.1 | |||
| AF305072.1 | AAG36977.1 | |||
| AF367023.1 | AAK53712.1 | |||
| AF367024.1 | AAK53713.1 | |||
| AF453246.1 | AAL86769.1 | |||
| AK289661.1 | ||||
| AK289922.1 | ||||
| AK290998.1 | ||||
| AK294188.1 | ||||
| AY029176.1 | AAK31594.1 | |||
| AY118170.1 | AAM81578.1 | |||
| BC032520.1 | AAH32520.1 | |||
| BX648990.1 | ||||
| DQ148487.1 | AAZ77804.1 | |||
| DQ148488.1 | AAZ77805.1 | |||
| DQ148489.1 | AAZ77806.1 | |||
| DQ148490.1 | AAZ77807.1 | |||
| DQ148491.1 | AAZ77808.1 | |||
| DQ148492.1 | AAZ77809.1 |
Target Gene Details
Entrez Gene ID: | 133015 |
Gene Name: | PARK2 coregulated like |
Gene Aliases: |
C4orf28 |
Location: |
Chr.4:20696282-20757766 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PACRGL | NR_133933.1 | |||
| XM_011513786.2 | 11 | 4112 | XP_011512088.1 | |
| XM_011513805.2 | 8 | 3653 | XP_011512107.1 | |
| XM_017007747.1 | 10 | 3867 | XP_016863236.1 | |
| XM_017007748.1 | 10 | 3818 | XP_016863237.1 | |
| XM_017007749.1 | 9 | 3733 | XP_016863238.1 | |
| XM_017007751.1 | 9 | 3935 | XP_016863240.1 | |
| XM_017007752.1 | 8 | 3782 | XP_016863241.1 | |
| XM_017007753.1 | 8 | 3652 | XP_016863242.1 | |
| XM_017007754.1 | 6 | 3709 | XP_016863243.1 | |
| XM_017007755.1 | 6 | 3526 | XP_016863244.1 | |
| XM_017007756.1 | 6 | 3519 | XP_016863245.1 | |
| XM_017007757.1 | 6 | 3455 | XP_016863246.1 | |
| XM_017007758.1 | 5 | 3404 | XP_016863247.1 | |
| XM_017007759.1 | 5 | 3352 | XP_016863248.1 | |
| AK098692.1 | BAC05381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829876 | Chr.4:20637113 - 20833998 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map