Assay Details
Target Gene Details
Entrez Gene ID: | 5428 |
Gene Name: | DNA polymerase gamma, catalytic subunit |
Gene Aliases: |
MDP1, MIRAS, MTDPS4A, MTDPS4B, PEO, POLG1, POLGA, SANDO, SCAE |
Location: |
Chr.15:89316305-89334795 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLG | NM_001126131.1 | NP_001119603.1 | ||
| NM_002693.2 | NP_002684.1 | |||
| AK291281.1 | ||||
| BC042571.1 | AAH42571.1 | |||
| BC050559.1 | AAH50559.1 | |||
| D84103.1 | BAA12223.1 | |||
| U60325.1 | AAC50712.1 | |||
| X98093.1 | CAA66719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038140 | Chr.15:89141274 - 89620108 on Build GRCh38 | Gain |
|
| nsv833086 | Chr.15:89275748 - 89437378 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map