Assay Details
Target Gene Details
Entrez Gene ID: | 101926987 |
Gene Name: | DLGAP4 antisense RNA 1 |
Gene Aliases: |
CCAT7 |
Location: |
Chr.20:36507702-36573275 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLGAP4-AS1 | NR_109939.1 |
Target Gene Details
Entrez Gene ID: | 10398 |
Gene Name: | myosin light chain 9 |
Gene Aliases: |
LC20, MLC-2C, MLC2, MRLC1, MYRL2 |
Location: |
Chr.20:36541484-36549823 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYL9 | NM_006097.4 | 4 | 881 | NP_006088.2 |
| NM_181526.2 | 3 | 719 | NP_852667.1 | |
| AF176042.1 | 3 | 817 | AAQ13653.1 | |
| AF176043.1 | 2 | 655 | AAQ13654.1 | |
| AK130879.1 | 4 | 846 | ||
| BC002648.2 | 3 | 658 | AAH02648.1 | |
| D82057.1 | 4 | 849 | BAB88917.1 | |
| J02854.1 | 3 | 819 | AAA59852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645738 | Chr.20:36548670 - 36555667 on Build GRCh38 | Loss |
|
| nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss |
|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map