Assay Details
Target Gene Details
Entrez Gene ID: | 55904 |
Gene Name: | lysine methyltransferase 2E |
Gene Aliases: |
HDCMC04P, MLL5, NKp44L |
Location: |
Chr.7:105014190-105114085 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KMT2E | NM_018682.3 | 26 | 4895 | NP_061152.3 |
| NM_182931.2 | 27 | 5043 | NP_891847.1 | |
| XM_005250493.1 | 26 | 4962 | XP_005250550.1 | |
| XM_011516400.1 | 28 | 5169 | XP_011514702.1 | |
| XM_017012435.1 | 27 | 5043 | XP_016867924.1 | |
| AF519459.1 | 26 | 4710 | AAM74947.1 | |
| AY147037.1 | 27 | 5043 | AAN17675.1 | |
| AY157990.1 | 24 | 4269 | AAN76325.1 | |
| AY222296.1 | AAO64395.1 | |||
| AY438698.1 | AAR13893.1 | |||
| AY438699.1 | AAR13894.1 |
Target Gene Details
Entrez Gene ID: | 6733 |
Gene Name: | SRSF protein kinase 2 |
Gene Aliases: |
SFRSK2 |
Location: |
Chr.7:105110708-105399366 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPK2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv237e55 | Chr.7:105097165 - 105512795 on Build GRCh38 | Gain |
|
| nsv608068 | Chr.7:104981955 - 105117036 on Build GRCh38 | Loss |
|
| nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map