Assay Details
Target Gene Details
Entrez Gene ID: | 11235 |
Gene Name: | programmed cell death 10 |
Gene Aliases: |
CCM3, TFAR15 |
Location: |
Chr.3:167683891-167735690 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDCD10 | NM_007217.3 | 9 | 1179 | NP_009148.2 |
NM_145859.1 | 9 | 1028 | NP_665858.1 | |
NM_145860.1 | 8 | 934 | NP_665859.1 | |
XM_005247086.4 | 9 | 1914 | XP_005247143.1 | |
XM_005247087.4 | 9 | 1919 | XP_005247144.1 | |
XM_005247088.3 | 8 | 1010 | XP_005247145.1 | |
XM_006713485.3 | 9 | 1074 | XP_006713548.1 | |
XM_011512368.2 | 10 | 1987 | XP_011510670.1 | |
XM_011512369.2 | 10 | 1992 | XP_011510671.1 | |
XM_017005644.1 | 9 | 1069 | XP_016861133.1 | |
XM_017005645.1 | 9 | 1910 | XP_016861134.1 | |
AF022385.1 | 8 | 934 | AAB72225.1 | |
AK291130.1 | ||||
BC002506.1 | 9 | 1191 | AAH02506.1 | |
BC016353.1 | 9 | 1032 | AAH16353.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
