Assay Details
Target Gene Details
Entrez Gene ID: | 7738 |
Gene Name: | zinc finger protein 184 |
Gene Aliases: |
kr-ZNF3 |
Location: |
Chr.6:27450742-27473118 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF184 | NM_001318891.1 | 6 | 1013 | NP_001305820.1 |
| NM_001318892.1 | 6 | 861 | NP_001305821.1 | |
| NM_001318893.1 | 6 | 1042 | NP_001305822.1 | |
| NM_007149.2 | 6 | 769 | NP_009080.2 | |
| XM_011514861.2 | 6 | 795 | XP_011513163.1 | |
| AK123011.1 | 5 | 1295 | ||
| AK131264.1 | 6 | 1021 | BAD18442.1 | |
| AK312804.1 | 6 | 1010 | ||
| BC022992.1 | 6 | 751 | AAH22992.1 | |
| U66561.1 | 3 | 409 | AAC51180.1 |
More Information
Set Membership: |
|

Genomic Map