Assay Details
Target Gene Details
Entrez Gene ID: | 57630 |
Gene Name: | SH3 domain containing ring finger 1 |
Gene Aliases: |
POSH, RNF142, SH3MD2 |
Location: |
Chr.4:169094256-169271098 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3RF1 | NM_020870.3 | 10 | 2420 | NP_065921.2 |
| AB040927.1 | 6 | 1335 | BAA96018.1 | |
| AK021429.1 | 10 | 2281 | BAB13822.1 | |
| AK025255.1 | 6 | 1252 | ||
| BC033203.1 | 9 | 1967 | ||
| BC041023.1 | 8 | 1603 | AAH41023.1 | |
| BC053671.1 | 8 | 2762 | AAH53671.1 |
More Information
Set Membership: |
|

Genomic Map