Assay Details
Target Gene Details
Entrez Gene ID: | 57467 |
Gene Name: | hedgehog acyltransferase-like |
Gene Aliases: |
C3orf3, GUP1, MBOAT3, MSTP002, OACT3 |
Location: |
Chr.3:42692663-42702849 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HHATL | NM_020707.3 | 10 | 1429 | NP_065758.3 |
| NR_027753.1 | 11 | 1314 | ||
| XM_006713274.2 | 10 | 1383 | XP_006713337.1 | |
| XM_006713275.2 | 9 | 1256 | XP_006713338.1 | |
| XM_011533969.2 | 11 | 1463 | XP_011532271.1 | |
| XM_011533970.2 | 10 | 1270 | XP_011532272.1 | |
| XM_017006935.1 | 11 | 1289 | XP_016862424.1 | |
| XM_017006936.1 | 11 | 1483 | XP_016862425.1 | |
| XM_017006937.1 | 10 | 1114 | XP_016862426.1 | |
| XM_017006938.1 | 9 | 1188 | XP_016862427.1 | |
| AB032999.1 | 5 | 654 | BAA86487.1 | |
| AB042554.1 | 10 | 1236 | BAB17758.1 | |
| AF109356.1 | 7 | 903 | AAQ13504.1 | |
| BC022529.1 | 11 | 1314 | AAH22529.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954472 | Chr.3:42683809 - 42713908 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map