Assay Details
Target Gene Details
Entrez Gene ID: | 84364 |
Gene Name: | ADP ribosylation factor GTPase activating protein 2 |
Gene Aliases: |
IRZ, NBLA10535, ZFP289, ZNF289 |
Location: |
Chr.11:47164298-47177503 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARFGAP2 | NM_001242832.1 | 14 | 1682 | NP_001229761.1 |
| NM_032389.4 | 15 | 1766 | NP_115765.2 | |
| XM_005253166.1 | 16 | 2261 | XP_005253223.1 | |
| XM_005253167.1 | 16 | 2228 | XP_005253224.1 | |
| XM_005253168.2 | 16 | 2186 | XP_005253225.1 | |
| XM_006718346.1 | 15 | 2219 | XP_006718409.1 | |
| XM_011520405.1 | 15 | 2102 | XP_011518707.1 | |
| XM_017018413.1 | 15 | 2186 | XP_016873902.1 | |
| XM_017018414.1 | 15 | 1582 | XP_016873903.1 | |
| XM_017018415.1 | 15 | 1534 | XP_016873904.1 | |
| AB073358.1 | 14 | 1376 | BAE45716.1 | |
| AK027482.1 | 15 | 1502 | BAB55144.1 | |
| AK098324.1 | 10 | 1112 | ||
| AK129511.1 | 5 | 459 | ||
| AK301785.1 | 14 | 1386 | ||
| AK304541.1 | 15 | 1341 | ||
| AK315992.1 | 9 | 1033 | ||
| AL834337.1 | 15 | 1485 | CAD39004.1 | |
| BC030148.2 | 15 | 1509 | AAH30148.1 |
More Information
Set Membership: |
|

Genomic Map