Assay Details
Target Gene Details
Entrez Gene ID: | 90957 |
Gene Name: | DEAH-box helicase 57 |
Gene Aliases: |
DDX57 |
Location: |
Chr.2:38797729-38875883 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX57 | NM_198963.1 | 23 | 4069 | NP_945314.1 |
| XM_011533154.2 | 23 | 4073 | XP_011531456.1 | |
| XM_011533155.2 | 23 | 4071 | XP_011531457.1 | |
| XM_011533156.2 | 23 | 4073 | XP_011531458.1 | |
| AB209249.1 | 17 | 2421 | BAD92486.1 | |
| AF070590.1 | 8 | 1082 | ||
| AF283512.1 | 19 | 2760 | AAM73547.1 | |
| BC033636.1 | 4 | 439 | AAH33636.1 | |
| BC053623.1 | 17 | 2370 | AAH53623.1 | |
| BC065278.1 | 18 | 2564 | AAH65278.1 | |
| BC131534.1 | 23 | 4041 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9646 | Chr.2:38131169 - 38855047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map