Assay Details
Target Gene Details
Entrez Gene ID: | 51531 |
Gene Name: | tRNA methyltransferase O |
Gene Aliases: |
C9orf156, HSPC219, NAP1 |
Location: |
Chr.9:97876321-97922570 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMO | NM_016481.4 | 4 | 957 | NP_057565.3 |
| XM_005252047.3 | 4 | 914 | XP_005252104.1 | |
| XM_005252048.4 | 5 | 1017 | XP_005252105.1 | |
| XM_005252049.4 | 3 | 739 | XP_005252106.1 | |
| XM_005252050.4 | 4 | 843 | XP_005252107.1 | |
| XM_011518776.2 | 4 | 914 | XP_011517078.1 | |
| XM_011518777.1 | 5 | 996 | XP_011517079.1 | |
| AF151053.1 | AAF36139.1 | |||
| AK000213.1 | 4 | 957 | BAA91013.1 | |
| AK023069.1 | 1 | 2326 | ||
| AK309270.1 | ||||
| AY189280.1 | 6 | 3011 | AAO86724.1 | |
| BC002863.2 | 4 | 937 | AAH02863.1 | |
| X86033.1 | 1 | 229 | CAA60025.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048135 | Chr.9:97904649 - 97954748 on Build GRCh38 | Loss |
|
| nsv525113 | Chr.9:97907517 - 97924533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map