Assay Details
Target Gene Details
Entrez Gene ID: | 3716 |
Gene Name: | Janus kinase 1 |
Gene Aliases: |
JAK1A, JAK1B, JTK3 |
Location: |
Chr.1:64833223-65067746 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JAK1 | NM_001320923.1 | 23 | 3249 | NP_001307852.1 |
| NM_001321852.1 | 22 | 3252 | NP_001308781.1 | |
| NM_001321853.1 | 24 | 3511 | NP_001308782.1 | |
| NM_001321854.1 | 23 | 3427 | NP_001308783.1 | |
| NM_001321855.1 | 23 | 3430 | NP_001308784.1 | |
| NM_001321856.1 | 22 | 3165 | NP_001308785.1 | |
| NM_001321857.1 | 22 | 3343 | NP_001308786.1 | |
| NM_002227.3 | 22 | 3346 | NP_002218.2 | |
| XM_017001212.1 | 22 | 3190 | XP_016856701.1 | |
| AB209057.1 | 21 | 3102 | BAD92294.1 | |
| AB219242.1 | 20 | 2993 | BAE02826.1 | |
| AK314257.1 | 23 | 3250 | ||
| BC132729.1 | 21 | 3064 | ||
| BX647675.1 | 8 | 950 | ||
| HQ628631.1 | 20 | 3026 | ||
| M64174.1 | 21 | 3071 | AAA36527.1 |
More Information
Set Membership: |
|

Genomic Map