Assay Details
Target Gene Details
Entrez Gene ID: | 245973 |
Gene Name: | ATPase H+ transporting V1 subunit C2 |
Gene Aliases: |
ATP6C2, VMA5 |
Location: |
Chr.2:10720974-10785110 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1C2 | NM_001039362.1 | NP_001034451.1 | ||
| NM_144583.3 | NP_653184.2 | |||
| XM_011510339.2 | XP_011508641.1 | |||
| XM_011510340.2 | XP_011508642.1 | |||
| XM_011510341.2 | XP_011508643.1 | |||
| XM_017003745.1 | XP_016859234.1 | |||
| AK292952.1 | ||||
| AY039759.1 | AAK83464.1 | |||
| BC012142.1 | AAH12142.1 | |||
| BX358022.2 | ||||
| CD636943.1 | ||||
| DB230703.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000731 | Chr.2:10618531 - 10803756 on Build GRCh38 | Gain |
|
| nsv833381 | Chr.2:10583339 - 10771158 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map