Assay Details
Target Gene Details
Entrez Gene ID: | 10451 |
Gene Name: | vav guanine nucleotide exchange factor 3 |
Gene Aliases: |
- |
Location: |
Chr.1:107571160-107964923 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VAV3 | NM_001079874.1 | 10 | 1935 | NP_001073343.1 |
| NM_006113.4 | 27 | 3596 | NP_006104.4 | |
| XM_005270360.2 | 26 | 3408 | XP_005270417.1 | |
| XM_017000053.1 | 25 | 3517 | XP_016855542.1 | |
| XM_017000055.1 | 26 | 3266 | XP_016855544.1 | |
| XM_017000056.1 | 16 | 2472 | XP_016855545.1 | |
| AF067817.1 | 27 | 3579 | AAC79695.1 | |
| AF118887.1 | 27 | 3588 | AAD20349.1 | |
| BC143969.1 | 28 | 3763 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830993 | Chr.1:107441433 - 107613979 on Build GRCh38 | Gain |
|
| dgv248n100 | Chr.1:107385172 - 108286989 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map